Assay Details
Target Gene Details
Entrez Gene ID: | 221191 |
Gene Name: | protease, serine 54 |
Gene Aliases: |
CT67, KLKBL4 |
Location: |
Chr.16:58279997-58295047 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS54 | NM_001080492.1 | NP_001073961.1 | ||
| NM_001305173.1 | NP_001292102.1 | |||
| NM_001305174.1 | NP_001292103.1 | |||
| AK058068.1 | BAB71649.1 | |||
| AK302175.1 | ||||
| AL137484.1 | CAB70765.1 | |||
| BC057843.1 |
More Information
Set Membership: |
|

Genomic Map