Assay Details
Target Gene Details
Entrez Gene ID: | 51451 |
Gene Name: | leucine carboxyl methyltransferase 1 |
Gene Aliases: |
CGI-68, LCMT, PPMT1 |
Location: |
Chr.16:25111709-25178231 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCMT1 | NM_001032391.1 | NP_001027563.1 | ||
| NM_016309.2 | NP_057393.2 | |||
| XM_005255354.3 | XP_005255411.1 | |||
| XM_011545862.2 | XP_011544164.1 | |||
| XM_011545864.1 | XP_011544166.1 | |||
| AF037601.1 | AAF18267.1 | |||
| AF151826.1 | AAD34063.1 | |||
| AK025884.1 | BAB15270.1 | |||
| AK223364.1 | BAD97084.1 | |||
| AK225271.1 | ||||
| AK291885.1 | ||||
| AK314409.1 | ||||
| AL137283.1 | CAB70677.1 | |||
| AL560574.3 | ||||
| BC001214.1 | AAH01214.1 | |||
| BC014217.2 | AAH14217.1 | |||
| BI459868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758638 | Chr.16:24932074 - 25175657 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map