Assay Details
Target Gene Details
Entrez Gene ID: | 102467146 |
Gene Name: | MAF transcriptional regulator RNA |
Gene Aliases: |
linc-MAF-4, lincMAF4 |
Location: |
Chr.16:79721312-79770532 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAFTRR | NR_104663.1 | |||
| BX108785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639248 | Chr.16:79709018 - 79770282 on Build GRCh38 | Loss |
|
| esv3572208 | Chr.16:79668676 - 80101641 on Build GRCh38 | Gain |
|
| nsv833296 | Chr.16:79658339 - 79855830 on Build GRCh38 | Loss |
|
| esv3582370 | Chr.16:79743720 - 79758473 on Build GRCh38 | Loss |
|
| nsv833297 | Chr.16:79698604 - 79886522 on Build GRCh38 | Gain |
|
| nsv827770 | Chr.16:79745778 - 79746591 on Build GRCh38 | Gain |
|
| nsv573285 | Chr.16:79124285 - 80093429 on Build GRCh38 | Gain |
|

Genomic Map