Assay Details
Target Gene Details
Entrez Gene ID: | 2734 |
Gene Name: | golgi glycoprotein 1 |
Gene Aliases: |
CFR-1, ESL-1, MG-160, MG160 |
Location: |
Chr.16:74447427-74607144 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLG1 | NM_001145666.1 | 26 | 7280 | NP_001139138.1 |
| NM_001145667.1 | 26 | 8352 | NP_001139139.1 | |
| NM_012201.5 | 27 | 7313 | NP_036333.2 | |
| NR_027264.1 | 27 | 8461 | ||
| NR_027265.1 | 21 | 7349 | ||
| AK057359.1 | 1 | 954 | ||
| BX648396.1 | 1 | 1488 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833278 | Chr.16:74382046 - 74550225 on Build GRCh38 | Loss |
|
| nsv1065964 | Chr.16:74054192 - 74541706 on Build GRCh38 | Gain |
|
| nsv833277 | Chr.16:74300889 - 74464235 on Build GRCh38 | Loss |
|
| esv2762054 | Chr.16:74303138 - 74473158 on Build GRCh38 | Gain |
|
| nsv1056566 | Chr.16:74422691 - 74448490 on Build GRCh38 | Loss |
|
| esv3639067 | Chr.16:74445112 - 74488116 on Build GRCh38 | Gain |
|
| esv3571587 | Chr.16:74432668 - 74469268 on Build GRCh38 | Gain |
|
| nsv509632 | Chr.16:74311644 - 74471617 on Build GRCh38 | Insertion |
|
| esv2758656 | Chr.16:74300882 - 74553287 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78102726] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map