Assay Details
Target Gene Details
Entrez Gene ID: | 23510 |
Gene Name: | potassium channel tetramerization domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.17:75032567-75065889 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD2 | NM_015353.2 | NP_056168.1 | ||
| NR_110834.1 | ||||
| NR_110835.1 | ||||
| XM_011524575.2 | XP_011522877.1 | |||
| AK307083.1 | ||||
| BC033329.1 | AAH33329.1 | |||
| D79998.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114786710] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map