Assay Details
Target Gene Details
Entrez Gene ID: | 30837 |
Gene Name: | suppressor of cytokine signaling 7 |
Gene Aliases: |
NAP4, NCKAP4 |
Location: |
Chr.17:38351876-38405593 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS7 | NM_014598.3 | NP_055413.1 | ||
| XM_017024551.1 | XP_016880040.1 | |||
| XM_017024552.1 | XP_016880041.1 | |||
| AB005216.1 | BAA22432.1 | |||
| BC128607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428341 | Chr.17:37855347 - 38600766 on Build GRCh38 | Gain+Loss |
|
| nsv2041 | Chr.17:38353353 - 38386215 on Build GRCh38 | Insertion |
|
| esv2758688 | Chr.17:37706608 - 38600766 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map