Assay Details
Target Gene Details
Entrez Gene ID: | 27077 |
Gene Name: | B9 domain containing 1 |
Gene Aliases: |
B9, EPPB9, MKS9, MKSR1 |
Location: |
Chr.17:19335372-19378193 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B9D1 | NM_001243473.2 | NP_001230402.1 | ||
| NM_001243475.2 | NP_001230404.1 | |||
| NM_001321214.1 | NP_001308143.1 | |||
| NM_001321215.1 | NP_001308144.1 | |||
| NM_001321217.1 | NP_001308146.1 | |||
| NM_001321218.1 | NP_001308147.1 | |||
| NM_001321219.1 | NP_001308148.1 | |||
| NM_015681.4 | NP_056496.1 | |||
| XM_005256607.2 | XP_005256664.1 | |||
| AB030506.1 | BAA82655.1 | |||
| AI300844.1 | ||||
| AK293815.1 | ||||
| AK297782.1 | ||||
| AK309320.1 | ||||
| BI833202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833393 | Chr.17:19146950 - 19361795 on Build GRCh38 | Gain+Loss |
|
| esv3582561 | Chr.17:19240831 - 19585268 on Build GRCh38 | Loss |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map