Assay Details
Target Gene Details
Entrez Gene ID: | 6397 |
Gene Name: | SEC14 like lipid binding 1 |
Gene Aliases: |
PRELID4A, SEC14L |
Location: |
Chr.17:77088643-77217101 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC14L1 | NM_001204408.1 | NP_001191337.1 | ||
| FM995495.1 | ||||
| FM995497.1 | ||||
| FM995498.1 | ||||
| FN357114.1 | ||||
| FN357120.1 | 1 | 346 |
Target Gene Details
Entrez Gene ID: | 654434 |
Gene Name: | small nucleolar RNA host gene 20 |
Gene Aliases: |
C17orf86, LINC00338, NCRNA00338, SCARNA16HG |
Location: |
Chr.17:77088643-77094986 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNHG20 | NR_027058.1 | |||
| BX648413.1 | ||||
| DA068554.1 | ||||
| DA916381.1 | ||||
| FN357113.1 | ||||
| FN357115.1 | ||||
| FN357116.1 | ||||
| FN357117.1 | ||||
| FN357118.1 | ||||
| FN357119.1 | ||||
| FN357121.1 | ||||
| FN357122.1 | ||||
| FN357123.1 | ||||
| FN357124.1 | ||||
| FN357125.1 | ||||
| FN357126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5784n54 | Chr.17:77049144 - 77101828 on Build GRCh38 | Gain |
|
| esv3641304 | Chr.17:77065524 - 77180984 on Build GRCh38 | Gain |
|
| esv3582914 | Chr.17:77086710 - 77091639 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs3214724] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map