Assay Details
Target Gene Details
Entrez Gene ID: | 3996 |
Gene Name: | LLGL1, scribble cell polarity complex component |
Gene Aliases: |
DLG4, HUGL, HUGL-1, HUGL1, LLGL, Lgl1, Mgl1 |
Location: |
Chr.17:18225593-18244875 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LLGL1 | NM_004140.3 | NP_004131.3 | ||
| XM_011523849.2 | XP_011522151.1 | |||
| XM_011523850.2 | XP_011522152.1 | |||
| XM_011523851.2 | XP_011522153.1 | |||
| XM_011523852.2 | XP_011522154.1 | |||
| XM_011523853.2 | XP_011522155.1 | |||
| XM_011523854.2 | XP_011522156.1 | |||
| XM_011523855.2 | XP_011522157.1 | |||
| XM_011523856.1 | XP_011522158.1 | |||
| BC028037.1 | AAH28037.1 | |||
| BC028214.1 | ||||
| BC151838.1 | ||||
| D50550.1 | BAA19516.1 | |||
| X86371.1 | CAA60130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9503 | Chr.17:18218160 - 18265852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map