Assay Details
Target Gene Details
Entrez Gene ID: | 10801 |
Gene Name: | septin 9 |
Gene Aliases: |
AF17q25, MSF, MSF1, NAPB, PNUTL4, SINT1, SeptD1 |
Location: |
Chr.17:77281410-77500596 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPT9 | NM_001113491.1 | NP_001106963.1 | ||
| NM_001293695.1 | NP_001280624.1 | |||
| XM_006721643.2 | XP_006721706.1 | |||
| AF189712.2 | AAF23373.1 | |||
| AF189713.3 | AAF23374.1 | |||
| AJ312323.1 | CAC42225.1 | |||
| AK300268.1 | ||||
| AK300270.1 | ||||
| BC054004.1 | AAH54004.1 | |||
| DC307336.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3284n100 | Chr.17:77118623 - 77628815 on Build GRCh38 | Gain |
|
| nsv833552 | Chr.17:77226556 - 77413897 on Build GRCh38 | Loss |
|
| esv34174 | Chr.17:77098835 - 77357976 on Build GRCh38 | Loss |
|
| esv2758704 | Chr.17:77130826 - 77299326 on Build GRCh38 | Gain |
|
| nsv833551 | Chr.17:77138921 - 77309684 on Build GRCh38 | Gain |
|
| nsv576077 | Chr.17:77264304 - 77297220 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map