Assay Details
Target Gene Details
Entrez Gene ID: | 84311 |
Gene Name: | mitochondrial ribosomal protein L45 |
Gene Aliases: |
L45mt, MRP-L45 |
Location: |
Chr.17:38297023-38323218 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL45 | NM_001278279.2 | NP_001265208.2 | ||
| NM_032351.5 | NP_115727.5 | |||
| AK172741.1 | BAD18730.1 | |||
| AK225031.1 | ||||
| AK293605.1 | ||||
| BC130382.1 | ||||
| BC130384.1 | ||||
| BG761720.1 | ||||
| BI767984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428341 | Chr.17:37855347 - 38600766 on Build GRCh38 | Gain+Loss |
|
| esv2758688 | Chr.17:37706608 - 38600766 on Build GRCh38 | Gain+Loss |
|
| nsv960482 | Chr.17:38263330 - 38317291 on Build GRCh38 | Duplication |
|
| esv33106 | Chr.17:38299424 - 38357921 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map