Assay Details
Target Gene Details
Entrez Gene ID: | 57674 |
Gene Name: | ring finger protein 213 |
Gene Aliases: |
ALO17, C17orf27, KIAA1618, MYMY2, MYSTR, NET57 |
Location: |
Chr.17:80260820-80398786 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF213 | NM_001256071.2 | NP_001243000.2 | ||
| NM_020954.3 | NP_066005.2 | |||
| XM_005257545.4 | XP_005257602.2 | |||
| XM_005257546.4 | XP_005257603.2 | |||
| XM_006721995.3 | XP_006722058.1 | |||
| XM_011525084.2 | XP_011523386.1 | |||
| XM_011525086.2 | XP_011523388.1 | |||
| XM_011525087.2 | XP_011523389.1 | |||
| AB537889.1 | ||||
| AF397204.1 | AAN63520.1 | |||
| AF397205.1 | AAN63521.1 | |||
| AL832920.1 | CAH10615.1 | |||
| BC036891.1 | AAH36891.1 | |||
| BC040341.1 | AAH40341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469591 | Chr.17:80085322 - 80293486 on Build GRCh38 | Loss |
|
| nsv1066976 | Chr.17:80256448 - 80351468 on Build GRCh38 | Gain |
|
| nsv576131 | Chr.17:80205094 - 80292960 on Build GRCh38 | Loss |
|
| esv3641371 | Chr.17:80277452 - 80297499 on Build GRCh38 | Loss |
|
| nsv833562 | Chr.17:80114523 - 80301339 on Build GRCh38 | Loss |
|
| esv3641370 | Chr.17:80274417 - 80303391 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115976016] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map