Assay Details
Target Gene Details
Entrez Gene ID: | 23510 |
Gene Name: | potassium channel tetramerization domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.17:75032567-75065889 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD2 | NM_015353.2 | NP_056168.1 | ||
| NR_110834.1 | ||||
| NR_110835.1 | ||||
| XM_011524575.2 | XP_011522877.1 | |||
| AK307083.1 | ||||
| BC033329.1 | AAH33329.1 | |||
| D79998.1 |
More Information
Set Membership: |
|

Genomic Map