Assay Details
Target Gene Details
Entrez Gene ID: | 5023 |
Gene Name: | purinergic receptor P2X 1 |
Gene Aliases: |
P2X1 |
Location: |
Chr.17:3896591-3916666 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX1 | NM_002558.3 | NP_002549.1 | ||
| XM_006721529.2 | XP_006721592.1 | |||
| XM_011523896.2 | XP_011522198.1 | |||
| XM_011523897.2 | XP_011522199.1 | |||
| XM_011523898.2 | XP_011522200.1 | |||
| XM_011523899.2 | XP_011522201.1 | |||
| XM_011523900.2 | XP_011522202.1 | |||
| AF020498.1 | AAC24494.1 | |||
| AK313264.1 | ||||
| BC027949.1 | AAH27949.1 | |||
| BC044657.1 | AAH44657.1 | |||
| U45448.1 | AAC50698.1 | |||
| X83688.1 | CAA58657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
|
| nsv833343 | Chr.17:3822173 - 3980223 on Build GRCh38 | Loss |
|
| esv2761940 | Chr.17:3767589 - 3997617 on Build GRCh38 | Gain |
|
| nsv524296 | Chr.17:3890009 - 3935332 on Build GRCh38 | Loss |
|
| nsv526126 | Chr.17:3860639 - 3993513 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map