Assay Details
Target Gene Details
Entrez Gene ID: | 6613 |
Gene Name: | small ubiquitin-like modifier 2 |
Gene Aliases: |
HSMT3, SMT3B, SMT3H2, SUMO3, Smt3A |
Location: |
Chr.17:75167730-75183003 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUMO2 | NM_001005849.1 | NP_001005849.1 | ||
| NM_006937.3 | NP_008868.3 | |||
| AK024823.1 | ||||
| AK225090.1 | ||||
| AK225263.1 | ||||
| AK311837.1 | ||||
| BC008450.1 | AAH08450.1 | |||
| BC016775.1 | AAH16775.1 | |||
| BC022340.1 | AAH22340.1 | |||
| BC062713.1 | AAH62713.1 | |||
| BC068465.1 | AAH68465.1 | |||
| BC070159.1 | AAH70159.1 | |||
| BC071645.1 | AAH71645.1 | |||
| BC071646.1 | AAH71646.1 | |||
| BC107853.1 | AAI07854.1 | |||
| BF700114.1 | ||||
| BG778261.1 | ||||
| L76416.1 | AAD45399.1 | |||
| X99585.1 | CAA67897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2141 | Chr.17:75172907 - 75196796 on Build GRCh38 | Deletion |
|
| nsv470602 | Chr.17:75090871 - 75208042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map