Assay Details
Target Gene Details
Entrez Gene ID: | 10014 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
HD5, NY-CO-9 |
Location: |
Chr.17:44076753-44123646 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC5 | NM_001015053.1 | NP_001015053.1 | ||
| NM_005474.4 | NP_005465.2 | |||
| XM_005256905.1 | XP_005256962.1 | |||
| XM_005256906.4 | XP_005256963.1 | |||
| XM_011524149.1 | XP_011522451.1 | |||
| XM_011524150.1 | XP_011522452.1 | |||
| XM_017023988.1 | XP_016879477.1 | |||
| XM_017023989.1 | XP_016879478.1 | |||
| XM_017023990.1 | XP_016879479.1 | |||
| XM_017023991.1 | XP_016879480.1 | |||
| XM_017023992.1 | XP_016879481.1 | |||
| XM_017023993.1 | XP_016879482.1 | |||
| XM_017023995.1 | XP_016879484.1 | |||
| XM_017023996.1 | XP_016879485.1 | |||
| AB011172.2 | BAA25526.2 | |||
| AK294760.1 | ||||
| BC051824.1 | AAH51824.1 | |||
| BM564861.1 | ||||
| CN290533.1 |
Target Gene Details
Entrez Gene ID: | 105371789 |
Gene Name: | uncharacterized LOC105371789 |
Gene Aliases: |
- |
Location: |
Chr.17:44115912-44120595 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371789 | NR_136415.1 | |||
| DB338921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833454 | Chr.17:43940848 - 44158311 on Build GRCh38 | Loss |
|
| nsv833455 | Chr.17:44108553 - 44297300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map