Assay Details
Target Gene Details
Entrez Gene ID: | 10476 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit D |
Gene Aliases: |
ATPQ |
Location: |
Chr.17:75038860-75046979 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5H | NM_001003785.1 | NP_001003785.1 | ||
| NM_006356.2 | NP_006347.1 | |||
| AA608885.1 | ||||
| AF061735.1 | AAG43146.1 | |||
| AF070650.1 | AAD20956.1 | |||
| AF086159.1 | ||||
| AF087135.1 | AAC36338.1 | |||
| AK095671.1 | 1 | 1005 | ||
| AK312230.1 | ||||
| BC029392.1 | ||||
| BC032245.1 | AAH32245.1 | |||
| BC038092.2 | AAH38092.1 |
Target Gene Details
Entrez Gene ID: | 23510 |
Gene Name: | potassium channel tetramerization domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.17:75032567-75065889 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD2 | NR_110835.1 | |||
| AK307083.1 | ||||
| AK311278.1 | ||||
| DA408645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893044 | Chr.17:75032657 - 75048093 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map