Assay Details
Target Gene Details
Entrez Gene ID: | 5345 |
Gene Name: | serpin family F member 2 |
Gene Aliases: |
A2AP, AAP, ALPHA-2-PI, API, PLI |
Location: |
Chr.17:1742808-1755265 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINF2 | NM_000934.3 | NP_000925.2 | ||
| NM_001165920.1 | NP_001159392.1 | |||
| NM_001165921.1 | NP_001159393.1 | |||
| XM_017024765.1 | XP_016880254.1 | |||
| AK303763.1 | ||||
| BC031592.1 | AAH31592.1 | |||
| BX107460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9490 | Chr.17:1653428 - 1828993 on Build GRCh38 | Loss |
|
| nsv1065585 | Chr.17:1733183 - 1918769 on Build GRCh38 | Gain |
|
| nsv516886 | Chr.17:1708905 - 1754359 on Build GRCh38 | Loss |
|
| nsv1946 | Chr.17:1717490 - 1781570 on Build GRCh38 | Insertion |
|
| nsv833335 | Chr.17:1713955 - 1827844 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map