Assay Details
Target Gene Details
Entrez Gene ID: | 124739 |
Gene Name: | ubiquitin specific peptidase 43 |
Gene Aliases: |
- |
Location: |
Chr.17:9643493-9729687 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP43 | NM_001267576.1 | 1 | 32 | NP_001254505.1 |
| NM_153210.4 | 1 | 32 | NP_694942.3 | |
| XM_011523639.2 | XP_011521941.1 | |||
| XM_011523641.1 | XP_011521943.1 | |||
| XM_011523642.2 | XP_011521944.1 | |||
| XM_017024159.1 | XP_016879648.1 | |||
| XM_017024162.1 | XP_016879651.1 | |||
| AK055188.1 | BAB70869.1 | |||
| BC136368.1 | 1 | 32 | ||
| BC144041.1 | 1 | 32 | ||
| BC171759.1 | 1 | 32 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833358 | Chr.17:9643743 - 9828246 on Build GRCh38 | Gain |
|
| nsv833357 | Chr.17:9518357 - 9696606 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map