Assay Details
Target Gene Details
Entrez Gene ID: | 22905 |
Gene Name: | epsin 2 |
Gene Aliases: |
EHB21 |
Location: |
Chr.17:19237377-19336715 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPN2 | NM_001102664.1 | NP_001096134.1 | ||
| NM_014964.4 | NP_055779.2 | |||
| NM_148921.3 | NP_683723.2 | |||
| AB028988.1 | ||||
| AF062084.1 | AAC78608.1 | |||
| AF062085.1 | AAC78609.1 | |||
| AK001491.1 | ||||
| AK001996.1 | ||||
| AK024115.1 | BAB14831.1 | |||
| AK092366.1 | ||||
| AK295652.1 | ||||
| AK298006.1 | ||||
| AK307455.1 | ||||
| BC070036.1 | AAH70036.1 | |||
| BC093972.1 | AAH93972.1 | |||
| BC093974.1 | AAH93974.1 | |||
| BC143264.1 | ||||
| DA220261.1 |
Target Gene Details
Entrez Gene ID: | 100874018 |
Gene Name: | EPN2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:19296596-19306261 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPN2-AS1 | NR_048576.1 | |||
| BG705989.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833393 | Chr.17:19146950 - 19361795 on Build GRCh38 | Gain+Loss |
|
| esv3582561 | Chr.17:19240831 - 19585268 on Build GRCh38 | Loss |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map