Assay Details
Target Gene Details
Entrez Gene ID: | 6117 |
Gene Name: | replication protein A1 |
Gene Aliases: |
HSSB, MST075, REPA1, RF-A, RP-A, RPA70 |
Location: |
Chr.17:1829979-1899554 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPA1 | NM_002945.3 | NP_002936.1 | ||
| AB209732.1 | BAD92969.1 | |||
| AK289704.1 | ||||
| BC018126.1 | AAH18126.1 | |||
| M63488.1 | AAA36584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33335 | Chr.17:1779713 - 2048672 on Build GRCh38 | Loss |
|
| nsv1065585 | Chr.17:1733183 - 1918769 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map