Assay Details
Target Gene Details
Entrez Gene ID: | 6875 |
Gene Name: | TATA-box binding protein associated factor 4b |
Gene Aliases: |
SPGF13, TAF2C2, TAFII105 |
Location: |
Chr.18:26226491-26391685 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF4B | NM_001293725.1 | NP_001280654.1 | ||
| NM_005640.2 | NP_005631.1 | |||
| NR_121653.1 | ||||
| XM_005258339.3 | XP_005258396.1 | |||
| XM_011526153.2 | XP_011524455.1 | |||
| XM_017025932.1 | XP_016881421.1 | |||
| XM_017025933.1 | XP_016881422.1 | |||
| AB234096.1 | ||||
| AY888048.1 | AAX14897.1 | |||
| AY888049.1 | AAX14898.1 | |||
| Y09321.1 | CAA70499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576576 | Chr.18:26159061 - 26340350 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72878294] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map