Assay Details
Target Gene Details
Entrez Gene ID: | 100505549 |
Gene Name: | uncharacterized LOC100505549 |
Gene Aliases: |
- |
Location: |
Chr.18:57630302-57669276 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100505549 | NM_001242804.1 | NP_001229733.1 | ||
DB059241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833663 | Chr.18:57549030 - 57718589 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv577032 | Chr.18:57517516 - 57756707 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv2751777 | Chr.18:57515803 - 57805084 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
