Assay Details
Target Gene Details
Entrez Gene ID: | 10666 |
Gene Name: | CD226 molecule |
Gene Aliases: |
DNAM-1, DNAM1, PTA1, TLiSA1 |
Location: |
Chr.18:69860228-69962086 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD226 | NM_001303618.1 | NP_001290547.1 | ||
| NM_001303619.1 | NP_001290548.1 | |||
| NM_006566.3 | NP_006557.2 | |||
| XM_005266642.3 | XP_005266699.1 | |||
| XM_005266643.3 | XP_005266700.1 | |||
| XM_006722374.3 | XP_006722437.1 | |||
| XM_017025524.1 | XP_016881013.1 | |||
| XM_017025525.1 | XP_016881014.1 | |||
| XM_017025526.1 | XP_016881015.1 | |||
| XM_017025527.1 | XP_016881016.1 | |||
| AJ537553.1 | CAD61184.1 | |||
| AK313199.1 | ||||
| BC074787.2 | AAH74787.1 | |||
| GU935335.1 | ||||
| GU935336.1 | ||||
| U56102.1 | AAC50560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32736 | Chr.18:69135149 - 71678611 on Build GRCh38 | Gain+Loss |
|
| esv3893135 | Chr.18:69877481 - 69889410 on Build GRCh38 | Loss |
|
| esv2717304 | Chr.18:69875904 - 69889715 on Build GRCh38 | Deletion |
|
| dgv632e214 | Chr.18:69875833 - 69891905 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map