Assay Details
Target Gene Details
Entrez Gene ID: | 5273 |
Gene Name: | serpin family B member 10 |
Gene Aliases: |
PI-10, PI10 |
Location: |
Chr.18:63907969-63936111 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB10 | NM_005024.2 | NP_005015.1 | ||
| XM_011526027.1 | XP_011524329.1 | |||
| XM_011526028.1 | XP_011524330.1 | |||
| XM_017025793.1 | XP_016881282.1 | |||
| XM_017025794.1 | XP_016881283.1 | |||
| AK314936.1 | ||||
| BC096217.3 | AAH96217.1 | |||
| BC096218.3 | AAH96218.1 | |||
| BC096219.3 | AAH96219.1 | |||
| BC096220.3 | AAH96220.1 | |||
| U35459.1 | AAC50282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain |
|
| nsv833673 | Chr.18:63897072 - 64097021 on Build GRCh38 | Gain |
|
| esv21739 | Chr.18:63715122 - 64278129 on Build GRCh38 | Gain |
|
| esv3642785 | Chr.18:63831255 - 63986077 on Build GRCh38 | Gain |
|
| nsv1067311 | Chr.18:63818011 - 66637685 on Build GRCh38 | Loss |
|
| nsv525889 | Chr.18:63845132 - 63948110 on Build GRCh38 | Loss |
|
| esv2758730 | Chr.18:63881807 - 63923851 on Build GRCh38 | Loss |
|
| esv3642788 | Chr.18:63922332 - 63954216 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73961814] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map