Assay Details
Target Gene Details
Entrez Gene ID: | 25941 |
Gene Name: | tubulin polyglutamylase complex subunit 2 |
Gene Aliases: |
C18orf10, HMFN0601, L17, PGs2 |
Location: |
Chr.18:36777647-36829393 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPGS2 | NM_001271956.1 | NP_001258885.1 | ||
| XM_017025700.1 | XP_016881189.1 | |||
| XM_017025701.1 | XP_016881190.1 | |||
| XM_017025702.1 | XP_016881191.1 | |||
| XM_017025703.1 | XP_016881192.1 | |||
| BM796279.1 | ||||
| DA512668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576651 | Chr.18:36017567 - 36959615 on Build GRCh38 | Loss |
|
| esv2758722 | Chr.18:36470072 - 36901904 on Build GRCh38 | Gain |
|
| nsv817789 | Chr.18:36536338 - 36856407 on Build GRCh38 | Gain |
|
| esv3642236 | Chr.18:36659865 - 36881208 on Build GRCh38 | Loss |
|
| esv3583016 | Chr.18:36337333 - 37122274 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map