Assay Details
Target Gene Details
Entrez Gene ID: | 101927275 |
Gene Name: | NDUFV2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:9121265-9136720 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFV2-AS1 | NR_110771.1 | 1 | 291 | |
| NR_110772.1 | 1 | 291 | ||
| BI520375.1 | 1 | 216 | ||
| HY015092.1 | 1 | 293 |

Genomic Map