Assay Details
Target Gene Details
Entrez Gene ID: | 9658 |
Gene Name: | zinc finger protein 516 |
Gene Aliases: |
HsT287 |
Location: |
Chr.18:76357682-76495255 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF516 | NM_014643.3 | NP_055458.1 | ||
| XM_011526269.1 | XP_011524571.1 | |||
| XM_011526270.2 | XP_011524572.1 | |||
| XM_011526271.2 | XP_011524573.1 | |||
| XM_011526272.2 | XP_011524574.1 | |||
| XM_011526273.2 | XP_011524575.1 | |||
| XM_011526274.2 | XP_011524576.1 | |||
| XM_011526275.2 | XP_011524577.1 | |||
| XM_011526276.1 | XP_011524578.1 | |||
| XM_011526277.2 | XP_011524579.1 | |||
| XM_017026097.1 | XP_016881586.1 | |||
| BC108287.1 | AAI08288.1 | |||
| D86975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2717388 | Chr.18:76288671 - 78160297 on Build GRCh38 | Deletion |
|
| nsv1063632 | Chr.18:76317786 - 77170410 on Build GRCh38 | Gain |
|
| esv3643163 | Chr.18:76213391 - 76437604 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map