Assay Details
Target Gene Details
Entrez Gene ID: | 642597 |
Gene Name: | A kinase (PRKA) anchor inhibitor 1 |
Gene Aliases: |
C18orf42 |
Location: |
Chr.18:5143673-5199997 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAIN1 | NM_001145194.1 | NP_001138666.1 | ||
| XM_011525728.2 | XP_011524030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066095 | Chr.18:4845604 - 5263909 on Build GRCh38 | Gain |
|
| dgv3317n100 | Chr.18:5012384 - 5236844 on Build GRCh38 | Gain |
|
| dgv184n21 | Chr.18:3526777 - 5930980 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map