Assay Details
Target Gene Details
Entrez Gene ID: | 346 |
Gene Name: | apolipoprotein C4 |
Gene Aliases: |
APO-CIV, APOC-IV |
Location: |
Chr.19:44942238-44945496 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOC4 | NM_001646.2 | NP_001637.1 | ||
| AV690709.1 | ||||
| BC020723.1 | AAH20723.1 | |||
| DB185657.1 | ||||
| EF371378.1 |
Target Gene Details
Entrez Gene ID: | 100533990 |
Gene Name: | APOC4-APOC2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.19:44942238-44949565 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOC4-APOC2 | NR_037932.1 | |||
| AK126257.1 |
More Information
Set Membership: |
|

Genomic Map