Assay Details
Target Gene Details
Entrez Gene ID: | 390940 |
Gene Name: | phospholipase A2 inhibitor and LY6/PLAUR domain containing |
Gene Aliases: |
- |
Location: |
Chr.19:43575399-43582104 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PINLYP | NM_001193621.1 | NP_001180550.1 | ||
| XM_011526970.2 | XP_011525272.1 | |||
| XM_011526971.2 | 1 | 224 | XP_011525273.1 | |
| DW009999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3603n100 | Chr.19:43304322 - 43590925 on Build GRCh38 | Gain |
|
| nsv1063149 | Chr.19:43554929 - 43583431 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map