Assay Details
Target Gene Details
Entrez Gene ID: | 23211 |
Gene Name: | zinc finger CCCH-type containing 4 |
Gene Aliases: |
C19orf7 |
Location: |
Chr.19:47064187-47114436 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC3H4 | NM_015168.1 | NP_055983.1 | ||
| XM_005258676.3 | XP_005258733.1 | |||
| XM_005258677.4 | XP_005258734.1 | |||
| XM_005258678.2 | XP_005258735.1 | |||
| XM_006723113.3 | XP_006723176.1 | |||
| XM_011526668.2 | XP_011524970.1 | |||
| XM_011526669.2 | XP_011524971.1 | |||
| XM_011526670.2 | XP_011524972.1 | |||
| XM_017026529.1 | XP_016882018.1 | |||
| XM_017026530.1 | XP_016882019.1 | |||
| XM_017026531.1 | XP_016882020.1 | |||
| AB028987.2 | ||||
| AK125279.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833851 | Chr.19:47058544 - 47208779 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map