Assay Details
Target Gene Details
Entrez Gene ID: | 56971 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 19 |
Gene Aliases: |
CEACM19, CEAL1 |
Location: |
Chr.19:44671452-44684355 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM19 | NM_001127893.1 | 1 | 184 | NP_001121365.1 |
| NM_020219.3 | 1 | 184 | NP_064604.2 | |
| BC083499.1 | 1 | 184 | AAH83499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062812 | Chr.19:44538275 - 44711525 on Build GRCh38 | Gain |
|
| nsv483005 | Chr.19:44530751 - 44705087 on Build GRCh38 | Loss |
|

Genomic Map