Assay Details
Target Gene Details
Entrez Gene ID: | 64377 |
Gene Name: | carbohydrate sulfotransferase 8 |
Gene Aliases: |
GALNAC4ST1, GalNAc4ST, PSS3 |
Location: |
Chr.19:33621955-33773509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHST8 | NM_001127895.1 | NP_001121367.1 | ||
| NM_001127896.1 | NP_001121368.1 | |||
| AB047801.1 | BAB19806.1 | |||
| BC011380.1 | AAH11380.1 | |||
| BC014250.2 | AAH14250.1 | |||
| BI917117.1 | ||||
| BX354555.2 | ||||
| DB500322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv139e55 | Chr.19:33406261 - 34032878 on Build GRCh38 | Gain |
|
| nsv509736 | Chr.19:33653214 - 33795128 on Build GRCh38 | Insertion |
|
| esv2758755 | Chr.19:33286168 - 34090656 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78506251] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map