Assay Details
Target Gene Details
Entrez Gene ID: | 147700 |
Gene Name: | kinesin light chain 3 |
Gene Aliases: |
KLC2, KLC2L, KLCt, KNS2B |
Location: |
Chr.19:45340740-45351520 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLC3 | NM_177417.2 | NP_803136.2 | ||
| BC062998.1 | ||||
| BC073841.1 | AAH73841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss |
|
| esv3644502 | Chr.19:45323147 - 45403933 on Build GRCh38 | Gain |
|
| dgv29n64 | Chr.19:45334011 - 45373709 on Build GRCh38 | Gain |
|
| esv3644503 | Chr.19:45335731 - 45345968 on Build GRCh38 | Loss |
|
| nsv579867 | Chr.19:45323108 - 45408744 on Build GRCh38 | Gain |
|
| esv3644504 | Chr.19:45342675 - 45373732 on Build GRCh38 | Gain |
|
| nsv1063547 | Chr.19:45322368 - 45400204 on Build GRCh38 | Gain |
|
| nsv833844 | Chr.19:45181051 - 45384148 on Build GRCh38 | Loss |
|
| nsv579868 | Chr.19:45334025 - 45359625 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73940011] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map