Assay Details
Target Gene Details
Entrez Gene ID: | 587 |
Gene Name: | branched chain amino acid transaminase 2 |
Gene Aliases: |
BCAM, BCATM, BCT2, PP18 |
Location: |
Chr.19:48795062-48811063 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCAT2 | NM_001164773.1 | NP_001158245.1 | ||
| NM_001190.3 | NP_001181.2 | |||
| NM_001284325.1 | 2 | 256 | NP_001271254.1 | |
| AF268047.1 | AAK38368.1 | |||
| AF268048.1 | AAK38369.1 | |||
| AK093627.1 | 2 | 233 | ||
| AK223522.1 | BAD97242.1 | |||
| AK309428.1 | ||||
| AK310101.1 | ||||
| AK310243.1 | 2 | 289 | ||
| AK314548.1 | ||||
| BC001900.1 | AAH01900.1 | |||
| BM802402.1 | 2 | 230 | ||
| DA129578.1 | ||||
| DC395685.1 | 2 | 289 | ||
| U68418.1 | AAB67672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833855 | Chr.19:48750098 - 48949781 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map