Assay Details
Target Gene Details
Entrez Gene ID: | 9466 |
Gene Name: | interleukin 27 receptor subunit alpha |
Gene Aliases: |
CRL1, IL-27RA, IL27R, TCCR, WSX1, zcytor1 |
Location: |
Chr.19:14031740-14052919 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL27RA | NM_004843.3 | NP_004834.1 | ||
| AF053004.1 | AAC39755.1 | |||
| AF106912.1 | AAG26090.1 | |||
| AF265242.1 | AAG27297.1 | |||
| AY358876.1 | AAQ89235.1 | |||
| BC028003.1 | AAH28003.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833761 | Chr.19:13998659 - 14136778 on Build GRCh38 | Loss |
|
| nsv578641 | Chr.19:14024296 - 14050933 on Build GRCh38 | Gain |
|
| nsv578640 | Chr.19:14019403 - 14042481 on Build GRCh38 | Gain |
|
| nsv578642 | Chr.19:14028192 - 14187630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map