Assay Details
Target Gene Details
Entrez Gene ID: | 3978 |
Gene Name: | DNA ligase 1 |
Gene Aliases: |
- |
Location: |
Chr.19:48115445-48170603 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIG1 | NM_000234.2 | NP_000225.1 | ||
| NM_001289063.1 | NP_001275992.1 | |||
| NM_001289064.1 | NP_001275993.1 | |||
| NM_001320970.1 | NP_001307899.1 | |||
| NM_001320971.1 | NP_001307900.1 | |||
| NR_110296.1 | ||||
| NR_135497.1 | ||||
| NR_135498.1 | ||||
| NR_135499.1 | ||||
| NR_135500.1 | ||||
| NR_135501.1 | ||||
| XM_017026809.1 | XP_016882298.1 | |||
| AB208791.1 | BAD92028.1 | |||
| AK225863.1 | ||||
| AK297296.1 | ||||
| AK300370.1 | ||||
| AK300607.1 | ||||
| AK314210.1 | ||||
| BC108318.1 | AAI08319.1 | |||
| BC110622.2 | AAI10623.1 | |||
| M36067.1 | AAA59518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833853 | Chr.19:47982539 - 48184317 on Build GRCh38 | Loss |
|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss |
|
| nsv953596 | Chr.19:48120744 - 48134943 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map