Assay Details
Target Gene Details
Entrez Gene ID: | 105221694 |
Gene Name: | BST2 interferon stimulated positive regulator (non-protein coding) |
Gene Aliases: |
lncBST2 |
Location: |
Chr.19:17405686-17415736 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BISPR | NR_130765.1 | |||
| NR_130766.1 | ||||
| AK055623.1 | ||||
| AK311380.1 | ||||
| DA383342.1 | ||||
| HY067963.1 |
Target Gene Details
Entrez Gene ID: | 93343 |
Gene Name: | multivesicular body subunit 12A |
Gene Aliases: |
CFBP, FAM125A |
Location: |
Chr.19:17405686-17425339 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MVB12A | NM_001304547.1 | NP_001291476.1 | ||
| BG684013.1 | ||||
| BY996463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833768 | Chr.19:17338840 - 17526738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map