Assay Details
Target Gene Details
Entrez Gene ID: | 56971 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 19 |
Gene Aliases: |
CEACM19, CEAL1 |
Location: |
Chr.19:44671452-44684355 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9737 | Chr.19:44674838 - 44703359 on Build GRCh38 | Gain |
|
| nsv1062812 | Chr.19:44538275 - 44711525 on Build GRCh38 | Gain |
|
| nsv483005 | Chr.19:44530751 - 44705087 on Build GRCh38 | Loss |
|

Genomic Map