Assay Details
Target Gene Details
Entrez Gene ID: | 342865 |
Gene Name: | V-set and transmembrane domain containing 2B |
Gene Aliases: |
- |
Location: |
Chr.19:29525431-29564555 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM2B | NM_001146339.1 | NP_001139811.1 | ||
| XM_005258856.3 | XP_005258913.1 | |||
| XM_011526902.2 | XP_011525204.1 | |||
| XM_011526903.2 | XP_011525205.1 | |||
| XM_011526904.2 | XP_011525206.1 | |||
| XM_011526905.2 | XP_011525207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833798 | Chr.19:29379175 - 29551378 on Build GRCh38 | Loss |
|
| nsv1061604 | Chr.19:28373028 - 29773397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map