Assay Details
Target Gene Details
Entrez Gene ID: | 10908 |
Gene Name: | patatin like phospholipase domain containing 6 |
Gene Aliases: |
BNHS, LNMS, NTE, NTEMND, OMCS, SPG39, iPLA2delta, sws |
Location: |
Chr.19:7534152-7561767 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA6 | NM_001166111.1 | NP_001159583.1 | ||
| NM_001166112.1 | NP_001159584.1 | |||
| NM_001166113.1 | NP_001159585.1 | |||
| NM_001166114.1 | NP_001159586.1 | |||
| NM_006702.4 | NP_006693.3 | |||
| AJ004832.1 | CAA06164.1 | |||
| AK293807.1 | ||||
| AK294021.1 | ||||
| AK302462.1 | ||||
| BC038229.1 | AAH38229.1 | |||
| BC050553.1 | AAH50553.1 | |||
| BC051768.1 | AAH51768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain |
|
| nsv833730 | Chr.19:7509505 - 7633597 on Build GRCh38 | Loss |
|
| esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map