Assay Details
Target Gene Details
Entrez Gene ID: | 54776 |
Gene Name: | protein phosphatase 1 regulatory subunit 12C |
Gene Aliases: |
LENG3, MBS85, p84, p85 |
Location: |
Chr.19:55090913-55117600 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R12C | NM_001271618.1 | NP_001258547.1 | ||
| NM_017607.3 | NP_060077.1 | |||
| XM_005259013.3 | XP_005259070.1 | |||
| XM_011527045.2 | XP_011525347.1 | |||
| AB209452.1 | BAD92689.1 | |||
| AF312028.1 | AAG60045.1 | |||
| AK027086.1 | BAB15651.1 | |||
| AK297187.1 | ||||
| AK297426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv817874 | Chr.19:55067334 - 55103555 on Build GRCh38 | Loss |
|
| esv3644829 | Chr.19:55030893 - 55138732 on Build GRCh38 | Gain |
|
| esv3644836 | Chr.19:55084362 - 55113486 on Build GRCh38 | Gain |
|
| esv34356 | Chr.19:55026094 - 55139319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map