Assay Details
Target Gene Details
Entrez Gene ID: | 5064 |
Gene Name: | paralemmin |
Gene Aliases: |
- |
Location: |
Chr.19:708953-748330 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALM | NM_001040134.1 | NP_001035224.1 | ||
| NM_002579.2 | NP_002570.2 | |||
| XM_005259565.4 | XP_005259622.1 | |||
| XM_005259566.4 | XP_005259623.1 | |||
| XM_017026850.1 | XP_016882339.1 | |||
| AK299809.1 | ||||
| AK315168.1 | ||||
| BC032449.1 | AAH32449.1 | |||
| DA521203.1 | ||||
| Y14770.1 | CAB37400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion |
|
| nsv577978 | Chr.19:294526 - 782854 on Build GRCh38 | Gain |
|
| nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion |
|
| nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117757692] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map