Assay Details
Target Gene Details
Entrez Gene ID: | 100507588 |
Gene Name: | transforming growth factor beta receptor 3 like |
Gene Aliases: |
TGFR-3L |
Location: |
Chr.19:7915211-7919097 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFBR3L | XM_011527610.2 | 1 | 186 | XP_011525912.1 |
| XM_011527611.2 | 1 | 186 | XP_011525913.1 | |
| XM_011527612.2 | 1 | 186 | XP_011525914.1 | |
| XM_011527613.2 | 1 | 186 | XP_011525915.1 | |
| XM_011527614.2 | 1 | 186 | XP_011525916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953961 | Chr.19:7881316 - 7955515 on Build GRCh38 | Deletion |
|
| nsv833733 | Chr.19:7799229 - 7995254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map