Assay Details
Target Gene Details
Entrez Gene ID: | 83546 |
Gene Name: | retbindin |
Gene Aliases: |
- |
Location: |
Chr.19:12825477-12835428 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTBDN | NM_001080997.2 | NP_001074466.1 | ||
| NM_001270440.1 | 7 | 1316 | NP_001257369.1 | |
| NM_001270441.1 | NP_001257370.1 | |||
| NM_001270442.1 | NP_001257371.1 | |||
| NM_001270443.1 | NP_001257372.1 | |||
| NM_001270444.1 | NP_001257373.1 | |||
| NM_001270445.1 | NP_001257374.1 | |||
| NM_031429.2 | NP_113617.1 | |||
| XM_005260089.1 | XP_005260146.1 | |||
| XM_005260090.3 | XP_005260147.1 | |||
| XM_011528343.1 | XP_011526645.1 | |||
| XM_011528344.2 | XP_011526646.1 | |||
| XM_011528345.1 | XP_011526647.1 | |||
| XM_017027343.1 | XP_016882832.1 | |||
| XM_017027344.1 | XP_016882833.1 | |||
| AB593122.1 | ||||
| AK093672.1 | 7 | 1316 | BAC04217.1 | |
| AY028917.1 | AAK26169.1 | |||
| BC005063.1 | AAH05063.1 | |||
| BG471112.1 | ||||
| BM688006.1 | ||||
| BM688127.1 | ||||
| BQ639040.1 | ||||
| BU689643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|

Genomic Map