Assay Details
Target Gene Details
Entrez Gene ID: | 80054 |
Gene Name: | CEBPA antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.19:33302857-33305057 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPA-AS1 | NR_026887.1 | 1 | 422 | |
| AK022417.1 | 1 | 323 | BAB14032.1 | |
| AL522369.3 | 1 | 422 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833808 | Chr.19:33130887 - 33350702 on Build GRCh38 | Loss |
|
| nsv2463 | Chr.19:33294939 - 33313582 on Build GRCh38 | Deletion |
|
| esv2758755 | Chr.19:33286168 - 34090656 on Build GRCh38 | Gain |
|

Genomic Map