Assay Details
Target Gene Details
Entrez Gene ID: | 90199 |
Gene Name: | WAP four-disulfide core domain 8 |
Gene Aliases: |
C20orf170, HEL-S-292, WAP8, dJ461P17.1 |
Location: |
Chr.20:45551152-45579326 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC8 | NM_130896.2 | NP_570966.2 | ||
| NM_181510.2 | NP_852611.2 | |||
| XM_017028119.1 | XP_016883608.1 | |||
| AF492015.1 | AAN70997.1 | |||
| AF492016.1 | AAN70998.1 | |||
| GQ472209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map