Assay Details
Target Gene Details
Entrez Gene ID: | 613266 |
Gene Name: | uncharacterized LOC613266 |
Gene Aliases: |
- |
Location: |
Chr.20:15892333-15985882 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC613266 | NR_130924.1 | |||
| DA380977.1 |
Target Gene Details
Entrez Gene ID: | 140733 |
Gene Name: | MACRO domain containing 2 |
Gene Aliases: |
C20orf133 |
Location: |
Chr.20:13995476-16053197 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MACROD2 | NM_001033087.1 | NP_001028259.1 | ||
| NM_080676.5 | NP_542407.2 | |||
| XM_017027675.1 | XP_016883164.1 | |||
| XM_017027676.1 | XP_016883165.1 | |||
| XM_017027677.1 | XP_016883166.1 | |||
| AK125899.1 | ||||
| AK131348.1 | BAD18504.1 | |||
| BC018687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057333 | Chr.20:15088323 - 16315570 on Build GRCh38 | Gain |
|
| nsv833929 | Chr.20:15784727 - 15996692 on Build GRCh38 | Gain |
|
| nsv3304 | Chr.20:15927189 - 15962052 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73900807] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map