Assay Details
Target Gene Details
Entrez Gene ID: | 149775 |
Gene Name: | GNAS antisense RNA 1 |
Gene Aliases: |
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG |
Location: |
Chr.20:58818918-58850903 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAS-AS1 | NR_002785.2 | |||
| AJ251759.1 | ||||
| BF589977.1 | ||||
| DB067618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955142 | Chr.20:58828346 - 58830845 on Build GRCh38 | Deletion |
|

Genomic Map